692026
2-Methylphenethyl bromide
97%
Synonym(s):
1-(2-Bromoethyl)-2-methylbenzene, 1-Bromo-2-o-tolylethane, 2-(2-Bromoethyl)toluene
About This Item
Assay
97%
form
liquid
refractive index
n20/D 1.5571
density
1.179 g/mL at 25 °C
SMILES string
Cc1ccccc1CCBr
InChI
1S/C9H11Br/c1-8-4-2-3-5-9(8)6-7-10/h2-5H,6-7H2,1H3
InChI key
PZXWZXNXBNIAHO-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
>230.0 °F
Flash Point(C)
> 110 °C
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service