Skip to Content
Merck
All Photos(1)

Key Documents

691062

Sigma-Aldrich

2-Methoxyethyl acetoacetate

Arxada quality, ≥97.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CO2CH2CH2OCH3
CAS Number:
Molecular Weight:
160.17
Beilstein:
2079850
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.5% (GC)

form

liquid

quality

Arxada quality

manufacturer/tradename

Arxada AG

impurities

≤0.10% water
≤1.5% 2-methoxyethanol

color

clear
yellowish

refractive index

n20/D 1.434 (lit.)

bp

120 °C/20 mmHg (lit.)

density

1.09 g/mL at 25 °C (lit.)

SMILES string

COCCOC(=O)CC(C)=O

InChI

1S/C7H12O4/c1-6(8)5-7(9)11-4-3-10-2/h3-5H2,1-2H3

InChI key

PLHCSZRZWOWUBW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 2 - STOT SE 3

Target Organs

Immune system, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service