Skip to Content
Merck
All Photos(1)

Documents

68730

Sigma-Aldrich

(4R,5S)-4-Methyl-5-phenyl-3-propionyl-2-oxazolidinone

≥97.0% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15NO3
CAS Number:
Molecular Weight:
233.26
Beilstein:
3549741
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥97.0% (sum of enantiomers, GC)

optical activity

[α]20/D +42±1°, c = 2% in methylene chloride

refractive index

n20/D 1.530

density

1.161 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CCC(=O)N1[C@H](C)[C@@H](OC1=O)c2ccccc2

InChI

1S/C13H15NO3/c1-3-11(15)14-9(2)12(17-13(14)16)10-7-5-4-6-8-10/h4-9,12H,3H2,1-2H3/t9-,12-/m1/s1

InChI key

ZMRFZBKROHCLIL-BXKDBHETSA-N

Other Notes

Chiral derivative of propionic acid used in highly enantioselective aldol, alkylation and acylation reactions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service