Skip to Content
Merck
All Photos(2)

Key Documents

685364

Sigma-Aldrich

Tetrakis[triphenylphosphine]palladium(0), ChemDose tablets

Loading: 2μmol per tablet

Synonym(s):

ChemDose, Tetrakis(triphenylphosphine)palladium(0) impregnated tablets, Palladium-tetrakis(triphenylphosphine) impregnated tablets

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd[(C6H5)3P]4
CAS Number:
Molecular Weight:
1155.56
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

tablet

matrix

Magnesium aluminometasilicate base material

storage temp.

2-8°C

SMILES string

[Pd].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9.c%10ccc(cc%10)P(c%11ccccc%11)c%12ccccc%12

InChI

1S/4C18H15P.Pd/c4*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;/h4*1-15H;

InChI key

NFHFRUOZVGFOOS-UHFFFAOYSA-N

Legal Information

ChemDose is a trademark of Reaxa Ltd.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenhan Zhang et al.
Angewandte Chemie (International ed. in English), 53(34), 8980-8984 (2014-07-01)
tert-Butoxyacetylene is shown to undergo Sonogashira coupling with aryl iodides to yield aryl-substituted tert-butyl ynol ethers. These intermediates participate in a [1,5]-hydride shift, which results in the extrusion of isobutylene and the generation of aryl ketenes. The ketenes are trapped
Chanchal Chakraborty et al.
ACS applied materials & interfaces, 7(34), 19034-19042 (2015-08-19)
A platinum(II)-based, luminescent, metallo-supramolecular polymer (PolyPtL1) having an inherent dipole moment was synthesized via complexation of Pt(II) ions with an asymmetric ligand L1, containing terpyridyl and pyridyl moieties. The synthesized ligand and polymer were well characterized by various NMR techniques
Debabrata Jana et al.
Organic & biomolecular chemistry, 13(43), 10663-10674 (2015-09-09)
Four pyrene-vinyl-tetraphenylethylene based conjugated materials were synthesized and characterized by FT-IR, NMR, and mass spectroscopy. The photophysical (including absorption, fluorescence, and fluorescence lifetime) and aggregation properties in tetrahydrofuran were investigated. The photophysical and aggregation behavior depends on the spacer, substituent
Shouchun Yin et al.
Soft matter, 11(22), 4424-4429 (2015-05-01)
In this paper, a pyrene moiety is incorporated into a bolaamphiphile to form a novel molecule denoted PRB. Above the critical micelle concentration, PRB forms nanodisks in the aqueous solution. The addition of acetate ions induces a morphological change in
Haiyin Li et al.
Macromolecular bioscience, 14(10), 1361-1367 (2014-06-14)
Phospholipid monomer and aggregation-induced emission (AIE) dye-based dimers are incorporated via reversible addition-fragmentation transfer polymerization to afford cross-linked zwitterionic fluorescent copolymers. Such copolymers are prone to self-assembly into fluorescent polymeric nanoparticles (FPNs) in physiological solution due to their amphiphilic nature.

Questions

1–7 of 7 Questions  
  1. Do the ChemDose® tablets disintegrate during the course of the reaction?

    1 answer
    1. They are normally recovered intact, however, they may disintegrate somewhat in aqueous reactions. Any disintegrated tablets can be removed from the reaction via filtration.

      Helpful?

  2. What is the pH stability of the ChemDose® tablets?

    1 answer
    1. The ChemDose® tablets are mildly basic and will dissolve in the presence of mineral acids.

      Helpful?

  3. Can I use ChemDose® tablets in a microwave or pressure vessel?

    1 answer
    1. Absolutely. Please see the Technical Information for Download section (below) for additional examples of cross-coupling reactions involving the use of microwaves.

      Helpful?

  4. How should I store ChemDose® tablets?

    1 answer
    1. As with most catalysts and ligands, as a general precaution it is best to store them in a refrigerator and/or under an inert atmosphere to maintain activity. Some catalysts, such as Pd(dppf)Cl2·CH2Cl2, are more robust and can be handled in air without issue. Please refer to the ChemDose® label for specific handling of a given catalyst, ligand, or reagent.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  6. Can I use ChemDose® tablets in any solvent?

    1 answer
    1. Yes, including aqueous solvents.

      Helpful?

  7. How should I dispose of spent ChemDose® tablets?

    1 answer
    1. They should be treated as solid waste, contaminated with reaction species. The matrix is non-toxic and environmentally benign.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service