Skip to Content
Merck
All Photos(1)

Key Documents

685321

Sigma-Aldrich

[6,6]-Phenyl-C61 butyric acid butyl ester

>97%

Synonym(s):

1-(3-Butoxycarbonylpropyl)-1-phenyl-[6.6]C61, PCBB, [60]PCB-C4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C75H20O2
CAS Number:
Molecular Weight:
952.96
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

description

functionalized fullerene

Assay

>97%

form

powder

storage temp.

2-8°C

SMILES string

CCCCOC(=O)CCCC2(c1ccccc1)C34c5c6c7c8c9c%10c(c%11c%12c3c%13c5c%14c%15c6c%16c7c%17c9c%18c%19c%10c%20c%11c%21c%12c%22c%13c%23c%14c%24c%15c%25c%16c%26c%17c%18c%27c%28c%19c%20c%29c%21c%30c%22c%23c%31c%24c%32c%25c%26c%27c%33c%28c%29c%30c%31c%32%33)C248

General description

[6,6]-Phenyl-C61 butyric acid butyl ester ([60]PCB-C4) is a fullerene derivative that is used as an acceptor molecule with good field effect mobility in organic electronics. It has a structural similarity to PCBM based fullerene and its aliphatic groups facilitate an increase in its solubility.[1][2]

Application

[60]PCB-C4 is a conducting polymer, which can be used in a variety of applications such as organic photovoltaic cells (OPVs) and bulk heterojunction (BHJ) solar cells.[2][3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystalline polymorphs of [6, 6]-phenyl-C61-butyric acid n-butyl ester (PCBNB)
Choi S, et al.
The Journal of Physical Chemistry B, 116(45), 13568-13574 (2012)
Polymer BHJ solar cell performance tuning by C60 fullerene derivative alkyl side-chain length
Kastner C, et al.
Journal of Polymer Science. Part B, Polymer Physics, 50(22), 1562-1566 (2012)
The Journal of Organic Chemistry, 60, 532-538 (1995)
MAPLE prepared heterostructures with oligoazomethine: Fullerene derivative mixed layer for photovoltaic applications
Stanculescu A, et al.
Applied Surface Science, 417, 183-195 (2017)
The Journal of Physical Chemistry B, 108, 11921-11926 (2004)

Articles

Progress in bulk heterojunction photodiodes since 1995 has improved device performance and widened research scope.

PCBM-based n-type semiconductors - Find p- and n-type organic semiconductors available with PCBM library & properties.

Find various photovoltaic and bioscience-based applications of fullerenes.

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service