All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H4ClN3
CAS Number:
Molecular Weight:
129.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
197-205 °C
functional group
chloro
SMILES string
Nc1ccnc(Cl)n1
InChI
1S/C4H4ClN3/c5-4-7-2-1-3(6)8-4/h1-2H,(H2,6,7,8)
InChI key
LPBDZVNGCNTELM-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service