Skip to Content
Merck
All Photos(1)

Documents

662259

Sigma-Aldrich

Ethyl 1,4-benzodioxan-2-carboxylate

97%

Synonym(s):

1,4-Benzodioxan-2-carboxylic acid, ethyl ester, Ethyl 2,3-dihydro-1,4-benzodioxin-2-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O4
Molecular Weight:
208.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.523

bp

88-95 °C/0.3 mmHg (lit.)

density

1.208 g/mL at 25 °C

SMILES string

CCOC(=O)C1COc2ccccc2O1

InChI

1S/C11H12O4/c1-2-13-11(12)10-7-14-8-5-3-4-6-9(8)15-10/h3-6,10H,2,7H2,1H3

InChI key

DDIGEMWIKJMEIU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service