Skip to Content
Merck
All Photos(1)

Key Documents

657611

Sigma-Aldrich

QuadraPure® AMPA

macroporous, 350-750 μm particle size

Synonym(s):

QuadraPure® Aminomethylphosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

product line

QuadraPure®

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: iron reactive
reagent type: catalyst
reagent type: chelator

particle size

350-750 μm

Application

Metal Scavenger: Fe, Cu, Ni, V, Al, Co

Legal Information

QuadraPure is a registered trademark of Johnson Matthey Finland Oy

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

QuadraPure Cartridges for Removal of Trace Metal from Reaction Mixtures in Flow
A. Hinchcliffe, et al.
Organic Process Research & Development, 11, 477-481 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service