Skip to Content
Merck
All Photos(1)

Key Documents

651672

Sigma-Aldrich

1-(3-Hydroxyphenyl) piperazine

97%

Synonym(s):

3-(1-Piperazino)phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O
CAS Number:
Molecular Weight:
178.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

217-221 °C (lit.)

SMILES string

Oc1cccc(c1)N2CCNCC2

InChI

1S/C10H14N2O/c13-10-3-1-2-9(8-10)12-6-4-11-5-7-12/h1-3,8,11,13H,4-7H2

InChI key

AYGYICRITMSJOC-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingguang Ren et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 249, 119299-119299 (2020-12-21)
Multiple types of metal ions and active small molecules (reactive nitrogen species, reactive oxygen species, reactive sulfur species, etc.) exist in living organisms. They have connections to each other and can interact and/or interfere with each other. To investigate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service