Skip to Content
Merck
All Photos(1)

Key Documents

647276

Sigma-Aldrich

4-Chlorobenzoin

97%

Synonym(s):

1-(4-Chlorophenyl)-2-hydroxy-2-phenylethanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4COCH(OH)C6H5
CAS Number:
Molecular Weight:
246.69
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

89-93 °C (lit.)

functional group

chloro
hydroxyl
ketone
phenyl

SMILES string

OC(c1ccccc1)C(=O)c2ccc(Cl)cc2

InChI

1S/C14H11ClO2/c15-12-8-6-11(7-9-12)14(17)13(16)10-4-2-1-3-5-10/h1-9,13,16H

InChI key

XYUKKJIDPMMCMJ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service