646946
6-Methoxy-2-naphthylmagnesium bromide solution
0.5 M in THF
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H9BrMgO
CAS Number:
Molecular Weight:
261.40
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
reaction suitability
reaction type: Grignard Reaction
concentration
0.5 M in THF
density
0.966 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
COc1ccc2cc([Mg]Br)ccc2c1
InChI
1S/C11H9O.BrH.Mg/c1-12-11-7-6-9-4-2-3-5-10(9)8-11;;/h3-8H,1H3;1H;/q;;+1/p-1
InChI key
SOGQGMFUEUMYJD-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Legal Information
Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-13.0 °F - closed cup
Flash Point(C)
-25 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service