Skip to Content
Merck
All Photos(1)

Key Documents

636711

Sigma-Aldrich

1,4-Bis(2-ethylhexyl)-2,5-diiodobenzene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H36I2
CAS Number:
Molecular Weight:
554.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

refractive index

n20/D 1.5564 (lit.)

density

1.3991 g/mL at 25 °C (lit.)

functional group

iodo

SMILES string

CCCCC(CC)Cc1cc(I)c(CC(CC)CCCC)cc1I

InChI

1S/C22H36I2/c1-5-9-11-17(7-3)13-19-15-22(24)20(16-21(19)23)14-18(8-4)12-10-6-2/h15-18H,5-14H2,1-4H3

InChI key

TYZQQLJIRWRBFL-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service