Skip to Content
Merck
All Photos(1)

Key Documents

62495

Sigma-Aldrich

Lithium diisopropylamide mono(tetrahydrofuran) complex solution

~1.5 M in cyclohexane, liquid, orange

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2NLi · C4H8O
CAS Number:
Molecular Weight:
179.23
Beilstein:
5894563
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

liquid

shelf life

limited shelf life

concentration

~1.5 M in cyclohexane

impurities

~10% ethylbenzene

color

orange

density

0.816 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

C1CCOC1.[Li]N(C(C)C)C(C)C

InChI

1S/C6H14N.C4H8O.Li/c1-5(2)7-6(3)4;1-2-4-5-3-1;/h5-6H,1-4H3;1-4H2;/q-1;;+1

InChI key

YTJXGDYAEOTOCG-UHFFFAOYSA-N

Caution

may become turbid on storage or may contain precipitate

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, hearing organs

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service