Skip to Content
Merck
All Photos(3)

Key Documents

590762

Sigma-Aldrich

5-(Trifluoromethyl)pyridine-2-thiol

97%

Synonym(s):

5-(Trifluoromethyl)-2-mercaptopyridine, 5-(Trifluoromethyl)-2-pyridinethiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4F3NS
CAS Number:
Molecular Weight:
179.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

154-159 °C (lit.)

functional group

fluoro

storage temp.

2-8°C

SMILES string

FC(F)(F)c1ccc(S)nc1

InChI

1S/C6H4F3NS/c7-6(8,9)4-1-2-5(11)10-3-4/h1-3H,(H,10,11)

InChI key

YFSCXLSUZZXNHW-UHFFFAOYSA-N

Application

5-(Trifluoromethyl)pyridine-2-thiol may be used to synthesize 5-trifluoromethylpyridine-2-sulfonyl chloride by reacting with chlorine gas in HCl solution.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Synthesis of [14C]-and [13C6]-labeled tipranavir and its potential hydroxyl metabolite and the glucuronide conjugate"
Latli B, et al.
Journal of Labelled Compounds & Radiopharmaceuticals, 51(08), 314- 320 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service