Skip to Content
Merck
All Photos(1)

Key Documents

578428

Sigma-Aldrich

(S)-(−)-5,5′,6,6′-Tetramethyl-3,3′-di-tert-butyl-1,1′-biphenyl-2,2′-diol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34O2
CAS Number:
Molecular Weight:
354.53
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Assay

97%

form

solid

optical activity

[α]20/D -83±3°, c = 0.4 in THF

mp

170-174 °C

SMILES string

Cc1cc(c(O)c(c1C)-c2c(C)c(C)cc(c2O)C(C)(C)C)C(C)(C)C

InChI

1S/C24H34O2/c1-13-11-17(23(5,6)7)21(25)19(15(13)3)20-16(4)14(2)12-18(22(20)26)24(8,9)10/h11-12,25-26H,1-10H3

InChI key

NMVVBVMYPLMIOU-UHFFFAOYSA-N

General description

(S)-5,5′,6,6′-Tetramethyl-3,3′-di-tert-butyl-1,1′-biphenyl-2,2′-diol, a biphenyl chiral ligand is used to prepare mono- and bidentate phosphites applicable as catalysts in the hydroformylation of allyl cyanide, heterocyclic olefins, and asymmetric hydrogenation of dimethyl itaconate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of new chiral monodentate phosphite ligands and their use in catalytic asymmetric hydrogenation
Hua Z, et al.
Organic Letters, 5(21), 3831-3834 (2003)
Synthesis and application of a new bisphosphite ligand collection for asymmetric hydroformylation of allyl cyanide
Cobley CJ, et al.
The Journal of Organic Chemistry, 69(12), 4031-4040 (2004)
Asymmetric Hydroformylation of Heterocyclic Olefins Mediated by Supramolecularly Regulated Rhodium-Bisphosphite Complexes
Rovira L, et al.
The Journal of Organic Chemistry, 80(20), 10397-10403 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service