Skip to Content
Merck
All Photos(1)

Key Documents

577936

Sigma-Aldrich

Bis(tert-butylimino)bis(dimethylamino)tungsten(VI)

97%, 99.99% trace metals basis

Synonym(s):

BTBMW

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((CH3)3CN)2W(N(CH3)2)2
CAS Number:
Molecular Weight:
414.23
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%
99.99% trace metals basis

form

liquid

reaction suitability

core: tungsten
reagent type: catalyst

bp

81 °C/0.02 mmHg (lit.)

density

1.305 g/mL at 25 °C (lit.)

SMILES string

CN(C)[W](=NC(C)(C)C)(=NC(C)(C)C)N(C)C

InChI

1S/2C4H9N.2C2H6N.W/c2*1-4(2,3)5;2*1-3-2;/h2*1-3H3;2*1-2H3;/q;;2*-1;+2

InChI key

JVCWKXBYGCJHDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

77.9 °F - closed cup - (own results)

Flash Point(C)

25.5 °C - closed cup - (own results)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service