Skip to Content
Merck
All Photos(1)

Key Documents

577812

Sigma-Aldrich

2,3′-Bithiophene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6S2
CAS Number:
Molecular Weight:
166.26
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

96%

mp

60-64 °C (lit.)

SMILES string

c1csc(c1)-c2ccsc2

InChI

1S/C8H6S2/c1-2-8(10-4-1)7-3-5-9-6-7/h1-6H

InChI key

ILQGHVXNYGLZSR-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcin Dąbrowski et al.
ACS applied materials & interfaces, 11(9), 9265-9276 (2019-02-05)
We present an improved approach for the preparation of highly selective and homogeneous molecular cavities in molecularly imprinted polymers (MIPs) via the combination of surface imprinting and semi-covalent imprinting. Toward that, first, a colloidal crystal mold was prepared via the

Articles

Professor Rivnay (Northwestern University, USA) discusses using organic mixed conductors as an alternative to efficiently bridge the ionic world of biology with contemporary microelectronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service