Skip to Content
Merck
All Photos(1)

Key Documents

563552

Sigma-Aldrich

2-(Trifluoromethyl)benzenesulfonamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4CF3SO2NH2
CAS Number:
Molecular Weight:
225.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

180-184 °C (lit.)

SMILES string

NS(=O)(=O)c1ccccc1C(F)(F)F

InChI

1S/C7H6F3NO2S/c8-7(9,10)5-3-1-2-4-6(5)14(11,12)13/h1-4H,(H2,11,12,13)

InChI key

AFFPZJFLSDVZBV-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service