Skip to Content
Merck
All Photos(1)

Key Documents

537764

Sigma-Aldrich

1-Hexyn-3-ol

90%, technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O
CAS Number:
Molecular Weight:
98.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

refractive index

n20/D 1.431

bp

118 °C

density

0.873 g/mL at 25 °C

SMILES string

CCCC(O)C#C

InChI

1S/C6H10O/c1-3-5-6(7)4-2/h2,6-7H,3,5H2,1H3

InChI key

LTFTWJYRQNTCHI-UHFFFAOYSA-N

General description

1-Hexyn-3-ol is a terminal propargylic alcohol. It undergoes microwave-accelerated coupling-isomerization reaction (MACIR) with (hetero)aryl halides to afford the corresponding enone.

Application

1-Hexyn-3-ol may be used in the synthesis of racemic 3-hydroxy-2-hexanone. It may also be used in the preparation of 1,2,3-triazoles.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

117.0 °F

Flash Point(C)

47.2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunfan Zou et al.
Journal of chemical ecology, 41(7), 670-677 (2015-07-08)
We report the identification of a novel pheromone structure from males of the cerambycid beetle Tylonotus bimaculatus Haldeman (Cerambycinae: Hesperophanini), a species native to eastern North America. Volatiles collected from adult males contained (2S,4E)-2-hydroxyoct-4-en-3-one (71%), (3R,4E)-3-hydroxyoct-4-en-2-one (15%), (E)-4-octen-2,3-dione (13%), and
Microwave-Accelerated Coupling-Isomerization Reaction (MACIR)?A General Coupling-Isomerization Synthesis of 1, 3-Diarylprop-2-en-1-ones.
Muller TJJ.
Advanced Synthesis & Catalysis, 348(18), 2565-2570 (2006)
Synthesis of New Carbohydrate Derivatives Via 1, 3-Dipolarcycloaddition Reaction.
Sharba AHK, et al.
Journal of Al-Nahrain University, 14(2), 9S-9S (2011)
The synthesis of 1, 1'-disubstituted bis-cyclopropanes by the reaction of substituted propargylic alcohols with CH 2 I 2?R 3 Al.
Ramazanov IR, et al.
Tetrahedron Letters, 50(29), 4233-4235 (2009)

Articles

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service