Skip to Content
Merck
All Photos(2)

Documents

536938

Sigma-Aldrich

Biphenyl-4-sulfonyl chloride

Synonym(s):

(4-Biphenylyl)sulfonyl chloride, 4-(Chlorosulfonyl)biphenyl, 4-Phenylbenzenesulfonic acid chloride, 4-Phenylbenzenesulfonyl chloride, p-Biphenylsulfonyl chloride, p-Phenylbenzenesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4SO2Cl
CAS Number:
Molecular Weight:
252.72
Beilstein:
2694373
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

103-108 °C (lit.)

SMILES string

ClS(=O)(=O)c1ccc(cc1)-c2ccccc2

InChI

1S/C12H9ClO2S/c13-16(14,15)12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9H

InChI key

ALBQXDHCMLLQMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of anilinopyrimidinesulfonamides as potential anticancer agents
  • Synthesis of inhibitors of the hypoxia inducible factor pathway
  • Preparation of hydroxy-sulfonyl-piperidinyl butyramides as HDAC inhibitors and potential antitumors
  • Palladium-catalyzed desulfitative C-arylation

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service