Skip to Content
Merck
All Photos(1)

Key Documents

529621

Sigma-Aldrich

Bis(triphenylphosphine)nickel(II) dichloride

99.99% trace metals basis

Synonym(s):

NiCl2(PPh3)2, Dichlorobis(triphenylphosphine)nickel(II), Nickel(II)bis(triphenylphosphine) dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)3P]2NiCl2
CAS Number:
Molecular Weight:
654.17
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

powder or crystals

reaction suitability

core: nickel

mp

250 °C (dec.) (lit.)

SMILES string

Cl[Ni]Cl.c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6

InChI

1S/2C18H15P.2ClH.Ni/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h2*1-15H;2*1H;/q;;;;+2/p-2

InChI key

ZBRJXVVKPBZPAN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Ni(PPh3)2Cl2 may be used to undertake nickel assisted phosphination of biaryl O,N triflates with chlorodiphenylphosphine.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chelating Retardation Effect in Nickel Assisted Phosphination: Syntheses of Atropisomeric P,N Ligands
Kwong FY, et al.
Tetrahedron, 56(45), 8893-8899 (2000)

Articles

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service