Skip to Content
Merck
All Photos(1)

Key Documents

525987

Sigma-Aldrich

N,N,5-Trimethylfurfurylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13NO
CAS Number:
Molecular Weight:
139.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.463 (lit.)

bp

171 °C (lit.)

density

0.917 g/mL at 25 °C (lit.)

SMILES string

CN(C)Cc1ccc(C)o1

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.0 °F - closed cup

Flash Point(C)

48.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J W Kosh et al.
Pharmaceutical research, 6(8), 709-713 (1989-08-01)
Famotidine and selected H2-antagonists were evaluated with respect to toxicity and selected pharmacological activities. When administered intraperitoneally to mice at a dose equivalent to 10 times their respective H2-antagonist ED50 values, no deaths were observed. Similarly, no alteration in brain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service