Skip to Content
Merck
All Photos(1)

Key Documents

525499

Sigma-Aldrich

2,5-Dibromo-3-butylthiophene

96%

Synonym(s):

2,5-Dibromo-3-octylthiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10Br2S
CAS Number:
Molecular Weight:
298.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

refractive index

n20/D 1.574 (lit.)

bp

270-281 °C (lit.)

density

1.67 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCc1cc(Br)sc1Br

InChI

1S/C8H10Br2S/c1-2-3-4-6-5-7(9)11-8(6)10/h5H,2-4H2,1H3

InChI key

YRSQCQUZWSQKKU-UHFFFAOYSA-N

Application

Conducting polymer precursor.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Oligothiophenes are important organic electronic materials which can be produced using synthetic intermediates and Suzuki coupling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service