Skip to Content
Merck
All Photos(1)

Key Documents

524824

Sigma-Aldrich

2,4,5-Trifluoro-3-methoxybenzoic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC6F3(OCH3)CO2H
CAS Number:
Molecular Weight:
206.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

105-112 °C (lit.)

SMILES string

COc1c(F)c(F)cc(C(O)=O)c1F

InChI

1S/C8H5F3O3/c1-14-7-5(10)3(8(12)13)2-4(9)6(7)11/h2H,1H3,(H,12,13)

InChI key

YVJHZWWMKFQKDC-UHFFFAOYSA-N

General description

2,4,5-Trifluoro-3-methoxybenzoic acid readily forms organotin(IV) complexes.[1]

Application

2,4,5-Trifluoro-3-methoxybenzoic acid may be used as a precursor for the preparation of quinolone derivatives. It may also be used to synthesize 1-(carboxymethyl)-6,7-difluoro-8-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid[2] and triaqua (1,10-phenanthroline-κ2N, N′)(2, 4, 5-trifluoro-3-methoxybenzoato-κO1) cobalt (II) 2, 4, 5-trifluoro-3-methoxybenzoate.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-pot synthesis and antimicrobial activity of novel quinolone heterocyclic derivatives.
Zheng H, et al.
Journal of Heterocyclic Chemistry, 47(6), 1411-1414 (2010)
Syntheses and crystal structures of di-and triorganotin (IV) derivatives with 2, 4, 5-trifluoro-3-methoxybenzoic acid.
Ma CL, et al.
Polyhedron, 25(17), 3405-3412 (2006)
Crystal structure of triaqua (1, 10-phenanthroline-?2N, N')(2, 4, 5-trifluoro-3-methoxybenzoato-?O1) cobalt (II) 2, 4, 5-trifluoro-3-methoxybenzoate.
Sun J.
Acta Crystallographica Section E, Structure Reports Online, 70(11), m367-m368 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service