Skip to Content
Merck
All Photos(1)

Documents

517674

Sigma-Aldrich

Praseodymium(III) acetylacetonate hydrate

≥99.9% trace metals basis

Synonym(s):

Pr(acac)3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pr(C5H7O2)3 · xH2O
CAS Number:
Molecular Weight:
438.23 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.9% trace metals basis

form

solid

reaction suitability

core: praseodymium
reagent type: catalyst

mp

130 °C (dec.) (lit.)

SMILES string

[H]O[H].CC(=O)\C=C(\C)O[Pr](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/3C5H8O2.H2O.Pr/c3*1-4(6)3-5(2)7;;/h3*3,6H,1-2H3;1H2;/q;;;;+3/p-3/b3*4-3-;;

InChI key

YOMFUQJQHBDYMC-KJVLTGTBSA-K

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service