Skip to Content
Merck
All Photos(1)

Documents

515353

Sigma-Aldrich

2,3-Dimethylbenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3CHO
CAS Number:
Molecular Weight:
134.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.553 (lit.)

bp

86-88 °C/10 mmHg (lit.)

density

1.029 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)c1cccc(C)c1C

InChI

1S/C9H10O/c1-7-4-3-5-9(6-10)8(7)2/h3-6H,1-2H3

InChI key

UIFVCPMLQXKEEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.1 °F - closed cup

Flash Point(C)

101.70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maximilian Ackermann et al.
Angiogenesis, 20(3), 359-372 (2017-03-12)
Nintedanib, a tyrosine kinase inhibitor approved for the treatment of idiopathic pulmonary fibrosis, has anti-fibrotic, anti-inflammatory, and anti-angiogenic activity. We explored the impact of nintedanib on microvascular architecture in a pulmonary fibrosis model. Lung fibrosis was induced in C57Bl/6 mice
Dustyn A Barnette et al.
Biochemical pharmacology, 170, 113661-113661 (2019-10-13)
Terbinafine N-dealkylation pathways result in formation of 6,6-dimethyl-2-hepten-4-ynal (TBF-A), a reactive allylic aldehyde, that may initiate idiosyncratic drug-induced liver toxicity. Previously, we reported on the importance of CYP2C19 and 3A4 as major contributors to TBF-A formation. In this study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service