511056
Acid Red 37 diammonium salt form
Dye content 70 %
About This Item
composition
Dye content, 70%
mp
>300 °C (lit.)
λmax
513 nm
λ
H2O reference
UV absorption
λ: 513 nm Amax: 0.00
SMILES string
N.N.CC(=O)Nc1ccc(\N=N\c2c(N)ccc3cc(cc(O)c23)S(O)(=O)=O)c(c1)S(O)(=O)=O
InChI
1S/C18H16N4O8S2.2H3N/c1-9(23)20-11-3-5-14(16(7-11)32(28,29)30)21-22-18-13(19)4-2-10-6-12(31(25,26)27)8-15(24)17(10)18;;/h2-8,24H,19H2,1H3,(H,20,23)(H,25,26,27)(H,28,29,30);2*1H3/b22-21+;;
InChI key
OFRRHUJAJNSLAT-ZPZFBZIMSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service