Skip to Content
Merck
All Photos(1)

Key Documents

484628

Sigma-Aldrich

Benztropine mesylate

95%

Synonym(s):

Benztropine methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25NO
CAS Number:
Molecular Weight:
307.43
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

95%

mp

135 °C (lit.)

SMILES string

CS(O)(=O)=O.CN1[C@H]2CC[C@@H]1C[C@@H](C2)OC(c3ccccc3)c4ccccc4

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clara Velázquez-Sánchez et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 34(12), 2497-2507 (2009-07-17)
Benztropine (BZT) analogs, a family of high-affinity dopamine transporter ligands, are molecules that exhibit pharmacological and behavioral characteristics predictive of significant therapeutic potential in cocaine addiction. Here, we examined in mice the effects of 3 alpha-[bis(4'-fluorophenyl)metoxy]-tropane (AHN-1055) on motor activity
Su-Min Li et al.
The Journal of pharmacology and experimental therapeutics, 336(2), 575-585 (2010-11-23)
Previous studies suggested that differences between the behavioral effects of cocaine and analogs of benztropine were related to the relatively slow onset of action of the latter compounds. Several N-substituted benztropine analogs with a relatively fast onset of effects were
Clara Velazquez-Sanchez et al.
The international journal of neuropsychopharmacology, 14(5), 655-665 (2010-08-26)
N-substituted benztropine (BZT) analogs are molecules that display high affinity for the dopamine transporter (DAT), therapeutic-like effects in animal models of cocaine abuse, and psychopharmacological characteristics consistent with those of a substitute medication for cocaine addiction. Since amphetamine (Amph) and
Antonio Ferragud et al.
Psychopharmacology, 207(2), 281-289 (2009-09-17)
N-substituted benztropine analogs are potent dopamine uptake inhibitors that display pharmacokinetic/dynamic properties consistent with the profile of a substitute medication for cocaine addiction. The purpose of the present experiments was to characterize in rats the addictive-like properties of one such
Brooke E Schmeichel et al.
Neuropharmacology, 64, 321-328 (2012-07-17)
Drugs used to treat attention deficit hyperactivity disorder (ADHD) improve prefrontal cortex (PFC)-dependent cognitive function. The majority of ADHD-related treatments act either as dual norepinephrine (NE) and dopamine (DA) reuptake inhibitors (psychostimulants) or selective NE reuptake inhibitors (SNRIs). Certain benztropine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service