Skip to Content
Merck
All Photos(1)

Key Documents

480223

Sigma-Aldrich

Poly(vinyl alcohol-co-vinyl acetate-co-itaconic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(OH)]x[CH2CH(O2CCH3)]y[CH2C(CO2H)(CH2CO2H)]Z
CAS Number:
MDL number:
UNSPSC Code:
12162002

mp

186-200 °C (lit.)

transition temp

Tg 79.9 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service