Skip to Content
Merck
All Photos(1)

Documents

479608

Sigma-Aldrich

Ethyl (S)-(−)-4-bromo-3-hydroxybutyrate

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH(OH)CH2CO2C2H5
CAS Number:
Molecular Weight:
211.05
Beilstein:
4657172
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

technical grade

optical activity

[α]22/D −11°, neat

refractive index

n20/D 1.474 (lit.)

bp

94-96 °C/2 mmHg (lit.)

density

1.468 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)C[C@H](O)CBr

InChI

1S/C6H11BrO3/c1-2-10-6(9)3-5(8)4-7/h5,8H,2-4H2,1H3/t5-/m0/s1

InChI key

AIZRKZQHJNWBEI-YFKPBYRVSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service