Skip to Content
Merck
All Photos(1)

Key Documents

478695

Sigma-Aldrich

4-Bromo-1H-imidazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3BrN2
CAS Number:
Molecular Weight:
146.97
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

131-135 °C (lit.)

functional group

bromo

SMILES string

Brc1c[nH]cn1

InChI

1S/C3H3BrN2/c4-3-1-5-2-6-3/h1-2H,(H,5,6)

InChI key

FHZALEJIENDROK-UHFFFAOYSA-N

General description

4-Bromo-1H-imidazole (4-Br-1H-Im) is a heterocyclic building block containing an imidazole ring. It crystallizes in the monoclinic space group P21/c.

Application

4-Bromo-1H-imidazole may be used as a starting material to prepare 5(4)-bromo-4(5)-nitroimidazole.
It may be used in the synthesis of the following aminoimidazoles via palladium catalyzed amination:
  • N-(4-ethoxyphenyl)-1H-imidazol-4-amine
  • N-(2,5-dimethoxyphenyl)-1H-imidazol-4-amine
  • N-(3-phenoxyphenyl)-1H-imidazol-4-amine
  • N-(4-cyanophenyl)-1H-imidazol-4-amine
  • N-(pyridin-3-yl)-1H-imidazol-4-amine

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-catalyzed amination of unprotected five-membered heterocyclic bromides.
Su M, et al.
Organic Letters, 16(3), 832-835 (2014)
Efficient synthesis of DNA containing the guanine oxidation-nitration product 5-guanidino-4-nitroimidazole: Generation by a postsynthetic substitution reaction.
Neeley WL, et al.
Organic Letters, 6(2), 245-248 (2004)
Crystal structure of 4-bromo-1H-imidazole, at 200 K, C3H3BrN2.
Hosten EC and Betz R.
Zeitschrift fur Kristallographie, 230(1), 27-28 (2015)
Guokai Cui et al.
Chemistry, an Asian journal, 12(21), 2863-2872 (2017-08-26)
A new strategy involving the computer-assisted design of substituted imidazolate-based ionic liquids (ILs) through tuning the absorption enthalpy as well as the basicity of the ILs to improve SO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service