Skip to Content
Merck
All Photos(1)

Key Documents

47479

Sigma-Aldrich

Fmoc-Trp-OPfp

≥98.0%

Synonym(s):

Fmoc-L-tryptophan pentafluorophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H21F5N2O4
CAS Number:
Molecular Weight:
592.51
Beilstein:
4228691
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0%

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

Fc1c(F)c(F)c(OC(=O)[C@H](Cc2c[nH]c3ccccc23)NC(=O)OCC4c5ccccc5-c6ccccc46)c(F)c1F

InChI

1S/C32H21F5N2O4/c33-25-26(34)28(36)30(29(37)27(25)35)43-31(40)24(13-16-14-38-23-12-6-5-7-17(16)23)39-32(41)42-15-22-20-10-3-1-8-18(20)19-9-2-4-11-21(19)22/h1-12,14,22,24,38H,13,15H2,(H,39,41)/t24-/m0/s1

InChI key

KLPGTAYCHANVFY-DEOSSOPVSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service