Skip to Content
Merck
All Photos(1)

Key Documents

473367

Sigma-Aldrich

(1S,4S)-(−)-2-(4-Fluorophenyl)-2,5-diazabicyclo[2.2.1]heptane hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13FN2 · HBr
CAS Number:
Molecular Weight:
273.14
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

optical activity

[α]22/D −68°, c = 1 in H2O

mp

225-230 °C (lit.)

SMILES string

Br.[H][C@]12CN(c3ccc(F)cc3)[C@]([H])(CN1)C2

InChI

1S/C11H13FN2.BrH/c12-8-1-3-10(4-2-8)14-7-9-5-11(14)6-13-9;/h1-4,9,11,13H,5-7H2;1H/t9-,11-;/m0./s1

InChI key

HDMUZFRUGPXOBN-ROLPUNSJSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service