Skip to Content
Merck
All Photos(1)

Key Documents

472883

Sigma-Aldrich

Tetra-O-acetyl-6,6′-di-O-(tert-butyldimethylsilyl)-D-lactal

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H56O13Si2
CAS Number:
Molecular Weight:
704.95
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

optical activity

[α]20/D −7°, c = 1.1 in chloroform

SMILES string

CC(=O)O[C@@H]1C=CO[C@H](CO[Si](C)(C)C(C)(C)C)[C@@H]1OC2O[C@H](CO[Si](C)(C)C(C)(C)C)[C@H](OC(C)=O)[C@H](OC(C)=O)[C@H]2OC(C)=O

Application

Important building block for both solution- and solid-phase synthesis of oligosaccharides.[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seeberger, P.H., et al.
Aldrichimica Acta, 30, 75-75 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service