Skip to Content
Merck
All Photos(1)

Documents

472786

Sigma-Aldrich

6-O-(tert-Butyldiphenylsilyl)-D-glucal

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28O4Si
CAS Number:
Molecular Weight:
384.54
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

optical activity

[α]25/D +9°, c = 1.1 in chloroform

impurities

2% ethyl acetate
3% dichloromethane

mp

−9 °C (lit.)

storage temp.

−20°C

SMILES string

CC(C)(C)[Si](OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O)(c1ccccc1)c2ccccc2

InChI

1S/C22H30O6Si/c1-22(2,3)29(16-10-6-4-7-11-16,17-12-8-5-9-13-17)28-15-19(25)21(27)20(26)18(24)14-23/h4-14,18-21,24-27H,15H2,1-3H3/t18-,19+,20+,21+/m0/s1

InChI key

BSEPYXNJJUUAEY-DOIPELPJSA-N

Application

Important building block for both solution- and solid-phase synthesis of oligosaccharides.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seeberger, P.H., et al.
Aldrichimica Acta, 30, 75-75 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service