Skip to Content
Merck
All Photos(1)

Key Documents

470724

Sigma-Aldrich

Antimony(III) propoxide

98%

Synonym(s):

Antimony tripropoxide, Tripropoxyantimony, Tripropoxystibine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sb(OC3H7)3
CAS Number:
Molecular Weight:
299.02
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

98%

form

liquid

reaction suitability

core: antimony
reagent type: catalyst

bp

83-85 °C/1 mmHg (lit.)

density

1.375 g/mL at 25 °C (lit.)

SMILES string

CCCO[Sb](OCCC)OCCC

InChI

1S/3C3H7O.Sb/c3*1-2-3-4;/h3*2-3H2,1H3;/q3*-1;+3

InChI key

CQOXUSHAXLMSEN-UHFFFAOYSA-N

Application

Precursor for amorphous antimonic acid films and powders used in electrical conductivity and morphology studies.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service