Skip to Content
Merck
All Photos(1)

Key Documents

462209

Sigma-Aldrich

2,3,9,10,16,17,23,24-Octakis(octyloxy)-29H,31H-phthalocyanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C96H146N8O8
CAS Number:
Molecular Weight:
1540.24
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

mp

>300 °C (lit.)

λmax

701 nm

SMILES string

CCCCCCCCOc1cc2c3nc(nc4[nH]c(nc5nc(nc6[nH]c(n3)c7cc(OCCCCCCCC)c(OCCCCCCCC)cc67)c8cc(OCCCCCCCC)c(OCCCCCCCC)cc58)c9cc(OCCCCCCCC)c(OCCCCCCCC)cc49)c2cc1OCCCCCCCC

InChI

1S/C96H146N8O8/c1-9-17-25-33-41-49-57-105-81-65-73-74(66-82(81)106-58-50-42-34-26-18-10-2)90-97-89(73)101-91-75-67-83(107-59-51-43-35-27-19-11-3)84(108-60-52-44-36-28-20-12-4)68-76(75)93(98-91)103-95-79-71-87(111-63-55-47-39-31-23-15-7)88(112-64-56-48-40-32-24-16-8)72-80(79)96(100-95)104-94-78-70-86(110-62-54-46-38-30-22-14-6)85(69-77(78)92(99-94)102-90)109-61-53-45-37-29-21-13-5/h65-72H,9-64H2,1-8H3,(H2,97,98,99,100,101,102,103,104)

InChI key

URYRXSHEJQXAAO-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service