Skip to Content
Merck
All Photos(1)

Key Documents

426970

Supelco

Poly(4-ethylstyrene-co-divinylbenzene)

mean pore size >75 Å, 500 μm mean particle size

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

composition

, ~80% (divinylbenzene)

mean particle size

500 μm

pore size

>1.75 mL/g pore volume
>75 Å mean pore size

density

0.54 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

General description

Contains no functional groups. Polymeric adsorbent for removing soluble organics from polar solvents. Fast adsorption/desorption kinetics for large organic molecules. Resists degradation up to 150°C. Methanol is a good regenerant.

Application

Insoluble polymeric adsorbent used to remove organic molecules from aqueous media.

Features and Benefits

Highly cross-linked polymer bead. Very versatile adsorbent, may have low capacity for smaller organic molecules.

Physical form

Polymeric adsorbent beads

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service