Skip to Content
Merck
All Photos(2)

Key Documents

425737

Sigma-Aldrich

Thulium(III) trifluoromethanesulfonate

98%

Synonym(s):

Thulium(III) triflate, Trifluoromethanesulfonic acid thulium(III) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3)3Tm
CAS Number:
Molecular Weight:
616.14
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

core: thulium
reagent type: catalyst

SMILES string

[Tm+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.Tm/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

PBASUZORNBYVFM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Asymmetric alpha-amination and cycloadditions
  • Benzyl etherification
  • Condensation of aldehydes, amines and vinyl ethers
  • Glycosylation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service