Skip to Content
Merck
All Photos(2)

Key Documents

423920

Sigma-Aldrich

1-Phenyl-1H-tetrazole-5-thiol sodium salt

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5N4NaS
CAS Number:
Molecular Weight:
200.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

>300 °C (lit.)

solubility

water: soluble 1g/15ml, clear

SMILES string

[Na]Sc1nnnn1-c2ccccc2

InChI

1S/C7H6N4S.Na/c12-7-8-9-10-11(7)6-4-2-1-3-5-6;/h1-5H,(H,8,10,12);/q;+1/p-1

InChI key

RSZMKAPXKXEWBY-UHFFFAOYSA-M

General description

1-Phenyl-1H-tetrazole-5-thiol sodium salt is a tetrazole derivative. The kinetics of its reaction with Diel′s-Alder adducts of 1,4-benzoquinone has been investigated.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics of the reaction of 1-phenyl-1H-tetrazole-5-thiol sodium salt with less substituted 1, 4-naphthoquinone 2, 3-epoxides.
O'Brien DF.
The Journal of Organic Chemistry, 33(1), 262-265 (1968)
Mohammad M Khodaei et al.
The Journal of organic chemistry, 73(7), 2527-2532 (2008-03-05)
In the presence of 1-phenyl-5-mercaptotetrazole as a nucleophile, electrochemical oxidations of 1,2- and 1,4-dihydroxybenzenes have been investigated in aqueous solution using cyclic voltammetry and controlled-potential coulometry. The voltammetric results indicate that an electrooxidative/Michael-type sequential reaction occurs between the mercaptide anion
J Kejha et al.
Ceskoslovenska farmacie, 39(7), 294-298 (1990-09-01)
For the investigation of new anti-inflammatory drugs, 1-phenyl-5-mercaptotetrazole (I) was selected as the principal structure. The purpose itself lay in an alkylation of the mercapto group with different remainders. In some cases at the same time a substituent was introduced
K Nesmĕrák et al.
Folia microbiologica, 45(2), 138-142 (2001-03-29)
The antifungal effect of substituted 1-phenyl-5-mercaptotetrazoles was tested with Candida tropicalis, C. pseudotropicalis, C. mogii, Trichosporon cutaneum, Cryptococcus albidus and S. cerevisiae. Candida strains exhibited the lowest sensitivity to the compounds; the most sensitive was S. cerevisiae. The MIC values
[Information from the Soviet Toxicology Center].
Gigiena truda i professional'nye zabolevaniia, (10)(10), 55-56 (1980-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service