Skip to Content
Merck
All Photos(1)

Key Documents

419923

Sigma-Aldrich

N-Boc-ethylenediamine

98%

Synonym(s):

N-Boc-1,2-diaminoethane, tert-Butyl N-(2-aminoethyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCONHCH2CH2NH2
CAS Number:
Molecular Weight:
160.21
Beilstein:
1932330
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.458 (lit.)

bp

72-80 °C/0.1 mmHg (lit.)

density

1.012 g/mL at 20 °C (lit.)

SMILES string

NCCNC(OC(C)(C)C)=O

InChI

1S/C7H16N2O2/c1-7(2,3)11-6(10)9-5-4-8/h4-5,8H2,1-3H3,(H,9,10)

InChI key

AOCSUUGBCMTKJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service