Skip to Content
Merck
All Photos(1)

Documents

416576

Sigma-Aldrich

3,5-Dichloro-4-fluorobenzotrifluoride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(F)CF3
CAS Number:
Molecular Weight:
232.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.462 (lit.)

bp

166-168 °C (lit.)

density

1.555 g/mL at 25 °C (lit.)

SMILES string

Fc1c(Cl)cc(cc1Cl)C(F)(F)F

InChI

1S/C7H2Cl2F4/c8-4-1-3(7(11,12)13)2-5(9)6(4)10/h1-2H

InChI key

BWQFQKZDLBJZAW-UHFFFAOYSA-N

General description

3,5-Dichloro-4-fluorobenzotrifluoride is an aryl fluorinated building block.

Application

3,5-Dichloro-4-fluorobenzotrifluoride may be used for the synthesis of 3-[4-[1-(2,6-dichloro-3-iodo-4-trifluoromethylphenyl)-5-iodopyrazolo]]-3-(trifluoromethyl)diazirine.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert E Sammelson et al.
The Journal of organic chemistry, 68(21), 8075-8079 (2003-10-11)
3-[4-[1-(2,6-dichloro-4-trifluoromethylphenyl)pyrazolo]]-3-(trifluoromethyl)diazirine is a fipronil-based (i.e. fiprole), high-affinity probe for the GABA receptor. For synthesis of the tritium-labeled version of this trifluoromethyldiazirinylfiprole ([(3)H]TDF) the required intermediate, 3-[4-[1-(2,6-dichloro-3-iodo-4-trifluoromethylphenyl)-5-iodopyrazolo]]-3-(trifluoromethyl)diazirine, was prepared in 10 steps from pyrazole and 3,5-dichloro-4-fluorobenzotrifluoride. One of the key transformations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service