Skip to Content
Merck
All Photos(1)

Key Documents

415294

Sigma-Aldrich

5-Iodo-1,3-dimethyluracil

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7IN2O2
CAS Number:
Molecular Weight:
266.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

225 °C (dec.) (lit.)

solubility

DMSO: soluble 10 mg/mL(lit.)

SMILES string

CN1C=C(I)C(=O)N(C)C1=O

InChI

1S/C6H7IN2O2/c1-8-3-4(7)5(10)9(2)6(8)11/h3H,1-2H3

InChI key

XUUVRIXNFFFPCM-UHFFFAOYSA-N

General description

The effect of 5-iodo-1,3-dimethyluracil on the incorporation of [125I] 5-iodo-2′-deoxyuridine [125I radiolabelled IdUrd] in DNA of cancer cells has been studied. Synthesis of 5-iodo-1,3-dimethyluracil from aromatic organomercurials; 5-(acetoxymercurio)-1,3-dimethyluracil or 5,5′-mercuriobis(1,3-dimethyluracil) has been reported.

Application

5-Iodo-1,3-dimethyluracil may be used for the synthesis of N-[2-[(1,3-dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)thio]-4-oxo[1]benzothieno[3,2-d]pyrimidin-3(4H)yl]methanesulfonamide.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yves M Dupertuis et al.
European journal of nuclear medicine and molecular imaging, 29(4), 499-505 (2002-03-27)
5-Iodo-2'-deoxyuridine (IdUrd), a thymidine (TdR) analogue, can be radiolabelled with iodine-125, an Auger radiation emitter, to provoke double-strand breaks once incorporated into DNA of cancer cells. We have previously shown that co-incubation of [125I]IdUrd with unlabelled IdUrd provided an additive
Organic iodine (I, III, and V) chemistry: 10 years of development at the Medical University of Warsaw, Poland.
Skulski L.
Molecules (Basel), 5(12), 1331-1371 (2000)
Mariarita Barone et al.
Molecular diversity, 17(3), 445-458 (2013-04-27)
The aim of this work was to evaluate the potential anti-inflammatory activity of eleven (5-15) new synthesized derivatives of benzo-thieno[3,2-d]pyrimidine on two cell models, namely human keratinocytes NCTC 2544 and mouse monocyte-macrophages J774. For the synthesis of test compounds an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service