Skip to Content
Merck
All Photos(1)

Key Documents

412589

Sigma-Aldrich

2-Amino-4-tert-amylphenol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(CH3)2C6H3(NH2)OH
CAS Number:
Molecular Weight:
179.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

Assay

97%

mp

119-120 °C (lit.)

SMILES string

CCC(C)(C)c1ccc(O)c(N)c1

InChI

1S/C11H17NO/c1-4-11(2,3)8-5-6-10(13)9(12)7-8/h5-7,13H,4,12H2,1-3H3

InChI key

APOFDNKYFPDSLE-UHFFFAOYSA-N

Application

2-Amino-4-tert-amylphenol was used in the synthesis of N1-(2-hydroxy-5-tert-amylphenyl)-1,4,5,8-naphthalenetetracarboxylic-1,8-anhydride-4,5-imide.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and properties of dimeric naphthalene diimides.
Gudeika D, et al.
Chemija, 23(3), 233-238 (2012)
Walton J Cabrera et al.
Journal of colloid and interface science, 265(1), 44-48 (2003-08-21)
The effect of phenols containing different electron-withdrawing substituents on the potentiometric responses of several liquid PVC membranes containing the complex trioctylmethylammonium-nitrate, TOMA(+)NO(3)(-), is analyzed. The results make it possible to separate these phenols into two groups; those phenols containing electron-releasing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service