Skip to Content
Merck
All Photos(1)

Documents

411558

Sigma-Aldrich

Lithium isopropoxide solution

1.0 M in hexanes

Synonym(s):

Lithium 2-propanolate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOLi
CAS Number:
Molecular Weight:
66.03
Beilstein:
3618614
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

form

liquid

reaction suitability

core: lithium

concentration

1.0 M in hexanes

bp

68-72 °C

density

0.69 g/mL at 25 °C

SMILES string

[Li+].CC(C)[O-]

InChI

1S/C3H7O.Li/c1-3(2)4;/h3H,1-2H3;/q-1;+1

InChI key

HAUKUGBTJXWQMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Self-heat. 1 - Skin Corr. 1B - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-16.6 °F - closed cup

Flash Point(C)

-27 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service