Skip to Content
Merck
All Photos(1)

Key Documents

410691

Sigma-Aldrich

tert-Butyl(dibromomethyl)dimethylsilane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CSi(CH3)2CHBr2
CAS Number:
Molecular Weight:
288.10
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.506 (lit.)

bp

60 °C/1 mmHg (lit.)

density

1.423 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)[Si](C)(C)C(Br)Br

InChI

1S/C7H16Br2Si/c1-7(2,3)10(4,5)6(8)9/h6H,1-5H3

InChI key

XINIZQRUNWPXNM-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service