Skip to Content
Merck
All Photos(1)

Documents

410403

Sigma-Aldrich

2-Hydrazino-4-(trifluoromethyl)pyrimidine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5F3N4
CAS Number:
Molecular Weight:
178.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

mp

99-101 °C (lit.)

SMILES string

NNc1nccc(n1)C(F)(F)F

InChI

1S/C5H5F3N4/c6-5(7,8)3-1-2-10-4(11-3)12-9/h1-2H,9H2,(H,10,11,12)

InChI key

XQCAINVUBSAUBJ-UHFFFAOYSA-N

Application

2-Hydrazino-4-(trifluoromethyl)pyrimidine may be used in the following studies:
  • As derivatization reagent for the simultaneous determination of testosterone (T) and dihydrotestosterone (DHT) in murine tissue and serum samples by liquid chromatography-electrospray ionization-tandem mass spectrometry.
  • Synthesis of organohydrazide chelate complexes of rhenium and technetium, having a metal-nitrogen-nitrogen linkage.
  • Preparation of 1-(4-methoxyphenyl)ethanone [(4-trifluoromethyl)pyrimidine-2-yl]hydrazone (NAS-75).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jukkarin Srivilai et al.
Steroids, 116, 67-75 (2016-10-31)
Steroid 5α-reductase (S5αR) plays an important role in metabolizing testosterone into active androgen dihydrotestosterone (DHT) which is involved in many androgen dependent disorders, such as androgenic alopecia, benign prostatic hyperplasia and acne. The method for screening for S5αR inhibition is
The synthesis and characterization of technetium and rhenium hydrazinopyrimidine chelate complexes.
Hirsch-Kuchma M, et al.
J. Chem. Soc., Dalton Trans., 18, 3185-3188 (1997)
Shailendra Kumar Sharma et al.
The Journal of biological chemistry, 278(46), 45661-45671 (2003-08-22)
The emergence of drug-resistant forms of Plasmodium falciparum emphasizes the need to develop new antimalarials. In this context, the fatty acid biosynthesis (FAS) pathway of the malarial parasite has recently received a lot of attention. Due to differences in the
Yan Weng et al.
Analytical biochemistry, 402(2), 121-128 (2010-04-07)
A novel method was established for simultaneous quantitation of testosterone (T) and dihydrotestosterone (DHT) in murine tissue and serum samples. Endogenous T and DHT, together with the internal standards 17alpha-methyl-T and 17alpha-methyl-DHT, were extracted from tissues and then derivatized by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service