Skip to Content
Merck
All Photos(1)

Key Documents

409235

Sigma-Aldrich

2-Methyl-1-(trimethylsilyloxy)-1-propene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHOSi(CH3)3
CAS Number:
Molecular Weight:
144.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

liquid

refractive index

n20/D 1.409 (lit.)

bp

114-116 °C/650 mmHg (lit.)

density

0.785 g/mL at 25 °C (lit.)

SMILES string

C\C(C)=C\O[Si](C)(C)C

InChI

1S/C7H16OSi/c1-7(2)6-8-9(3,4)5/h6H,1-5H3

InChI key

KZVPFSJPLBOVLO-UHFFFAOYSA-N

General description

2-Methyl-1-(trimethylsilyloxy)-1-propene is an enol ether.

Application

2-Methyl-1-(trimethylsilyloxy)-1-propene may be used as reagent in the direct conversion of α-amino acids into β-amino aldehydes.[1]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos J Saavedra et al.
Organic & biomolecular chemistry, 10(22), 4448-4461 (2012-05-05)
A direct method for the transformation of α-amino acids into β-amino aldehydes was developed, and applied to the modification of the C-terminal residue of peptides. The method takes place in good yields and under mild conditions. The application of this
L Nassi et al.
Mutation research, 198(1), 53-60 (1988-03-01)
Generation of triplet ketones, either chemically through thermal decomposition of 3-hydroxymethyl-3,4,4-trimethyl-1,2-dioxetane and 3-[N-(pyridino)carbamoyl]methyl-3,4,4-trimethyl-1,2-dioxetane++ + or enzymatically via the aerobic oxidation of isobutyraldehyde trimethylsilyl enol ether catalyzed by horse-radish peroxidase, triggers the SOS function sfiA in E. coli. Although the observed
G Cilento et al.
Toxicology letters, 67(1-3), 17-28 (1993-04-01)
Selected enzymatic systems can efficiently produce a product in the electronically excited triplet state. Earlier, only the formation of electronically excited singlet species was known. The formation of triplet species has been demonstrated with both normal substrates/metabolites and with xenobiotics

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service