Skip to Content
Merck
All Photos(1)

Documents

40702

Sigma-Aldrich

5,7-Dimethyl-8-quinolinol

≥98.0% (GC)

Synonym(s):

5,7-Dimethyl-8-hydroxyquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO
CAS Number:
Molecular Weight:
173.21
Beilstein:
1451007
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.0% (GC)

mp

95-98 °C

SMILES string

Cc1cc(C)c2cccnc2c1O

InChI

1S/C11H11NO/c1-7-6-8(2)11(13)10-9(7)4-3-5-12-10/h3-6,13H,1-2H3

InChI key

UKWMNFHGYXRSFQ-UHFFFAOYSA-N

General description

5,7-Dimethyl-8-quinolinol (Me2-quoH) is a specific non-extracting reagent for lanthanide ions at pH below 11. It is useful for the extraction of Al3+ metal ions and the transition metal ions at ordinary pH conditions.

Application

5,7-Dimethyl-8-quinolinol has been used in the preparation of the following:
  • tetrakis(5,7-dimethylquinolin-8-olato-κ2N,O)zirconium(IV) dimethylformamide disolvate
  • tetrakis (5, 7-dimethylquinolin-8-olato-κ2N,O) hafnium (IV) dimethylformamide disolvate
  • [Ru(Me2-quo)(dcbpy)2]+Cl- (dcbpy = 2,2′-bipyridinyl-4,4′-dicarboxyl acid; Me2-quo = 5,7-dimethyl-8-hydroxyquinolinate)
  • bis(5,7-dimethyl-8-hydroxyquinolinato)platinum(II) complex

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bis (5,7-dimethyl-8-hydroxyquinolinato) platinum (II) Complex for Efficient Organic Heterojunction Solar Cells.
Low KH, et al.
Chemistry - An Asian Journal, 6(12), 3223-3229 (2011)
Tetrakis (5, 7-dimethylquinolin-8-olato-κ2N,O) zirconium (IV) dimethylformamide disolvate.
Steyn M, et al.
Acta Crystallographica Section E, Structure Reports Online, 68(11), 1344-1345 (2012)
Tetrakis (5, 7-dimethylquinolin-8-olato-κ2N,O) hafnium (IV) dimethylformamide disolvate.
Viljoen J, et al.
Acta Crystallographica Section E, Structure Reports Online, 67(10), 1428-1429 (2011)
Ruthenium oxyquinolate complexes for dye-sensitized solar cells.
Dragonetti C, et al.
Inorgorganica Chimica Acta, 405, 98-104 (2013)
5, 7-Dimethyl-8-quinolinol as a Specific Non-extracting Reagent for Lanthanide Ions at Usual pH Region.
Takayanagi T, et al.
Chemistry Letters (Jpn), 4, 687-690 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service