Skip to Content
Merck
All Photos(1)

Key Documents

405930

Sigma-Aldrich

1,3-Bis(isocyanatomethyl)benzene

99%

Synonym(s):

m-Xylylene diisocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH2NCO)2
CAS Number:
Molecular Weight:
188.18
Beilstein:
781825
EC Number:
MDL number:
UNSPSC Code:
12162002

Assay

99%

refractive index

n20/D 1.591 (lit.)

bp

130 °C/2 mmHg (lit.)

mp

−7 °C (lit.)

density

1.202 g/mL at 25 °C (lit.)

SMILES string

O=C=NCc1cccc(CN=C=O)c1

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiyuki Ohtake et al.
Toxicology, 393, 9-14 (2017-11-05)
The skin sensitization potential of chemicals has been traditionally assessed using regulatory accepted in vivo methods, such as guinea pig maximization test or mouse local lymph node assays (LLNAs). A huge effort to reduce and replace the use of animals

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service